missing translation for 'onlineSavingsMsg'
Learn More

Niobium slug, 6.35mm (0.25in) dia x 12.7mm (0.50in) length, Puratronic™, 99.99% (metals basis excluding Ta), Ta 300ppm

Catalog No. AA35365QJ
Change view
Click to view available options
Quantity:
1 Slug
5 Slugs
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35365QJ 5 Slugs
AA35365QI 1 Slug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA35365QJ Supplier Thermo Scientific Chemicals Supplier No. 035365QJ

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-03-1 | Nb | 92.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3-1-7440
Molecular Formula Nb
Molecular Weight (g/mol) 92.91
MDL Number MFCD00011126
InChI Key GUCVJGMIXFAOAE-UHFFFAOYSA-N
Synonym columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
PubChem CID 23936
ChEBI CHEBI:33344
IUPAC Name niobium
SMILES [Nb]

Specifications

Quantity 5 Slugs
Odor Odorless
Weight ≈3.4g/slug
Form Slug
Percent Purity 99.99%
Grade Puratronic
Assay (metals basis excluding Ta), Ta 300ppm
Chemical Name or Material Niobium slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length
EINECSNumber 231-113-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only