missing translation for 'onlineSavingsMsg'
Learn More

Niobium silicide, 99.85% (metals basis)

CAS: 12034-80-9 | NbSi2 | 149.076 g/mol

$808.51 - $808.51

Chemical Identifiers

CAS 12034-80-9
Molecular Formula NbSi2
Molecular Weight (g/mol) 149.076
MDL Number MFCD00151537
InChI Key GXAAOBARRLPQRY-UHFFFAOYSA-N
Synonym niobium silicide, niobium disilicide, bis $l^ 2, columbium silicide,
PubChem CID 6336884
IUPAC Name bis($l^{2}-silanylidene)niobium
SMILES [Si]=[Nb]=[Si]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1310022
View Documents
Thermo Scientific Chemicals
01310022
100 g
Each for $808.51
Only null left
 
Description

Description

Niobium silicide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12034-80-9
149.076
GXAAOBARRLPQRY-UHFFFAOYSA-N
6336884
[Si]=[Nb]=[Si]
NbSi2
MFCD00151537
niobium silicide, niobium disilicide, bis $l^ 2, columbium silicide,
bis($l^{2}-silanylidene)niobium

Specifications

1950°C
Powder
99.85% (metals basis)
MFCD00151537
Insoluble in water.
[Si]=[Nb]=[Si]
149.076
149.08
5.7
12034-80-9
100 g
NbSi2
niobium silicide, niobium disilicide, bis $l^ 2, columbium silicide,
GXAAOBARRLPQRY-UHFFFAOYSA-N
bis($l^{2}-silanylidene)niobium
6336884
Powder
Niobium silicide
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c

H319-H335

EINECSNumber : 234-812-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only