missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Niobium rod, 6.35mm (0.25in) dia, annealed, 99.8% (metals basis)
CAS: 7440-03-1 | Nb | 92.91 g/mol
Supplier: Thermo Scientific Chemicals 010273BU
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Niobium rod | |
| 3-1-7440 | |
| Odorless | |
| Rod | |
| Nb | |
| columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio | |
| [Nb] | |
| 92.91 | |
| CHEBI:33344 |
| 6.35mm (0.25 in.) dia., Annealed, Temper: soft | |
| 25 cm | |
| ≈2.7g/cm | |
| (metals basis) | |
| MFCD00011126 | |
| GUCVJGMIXFAOAE-UHFFFAOYSA-N | |
| niobium | |
| 23936 | |
| 99.8% |
Chemical Identifiers
| 3-1-7440 | |
| 92.91 | |
| GUCVJGMIXFAOAE-UHFFFAOYSA-N | |
| 23936 | |
| niobium |
| Nb | |
| MFCD00011126 | |
| columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio | |
| CHEBI:33344 | |
| [Nb] |
Safety and Handling
EINECSNumber : 231-113-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only