missing translation for 'onlineSavingsMsg'
Learn More

Niobium powder, APS 1-5 micron, 99.8% (metals basis)

CAS: 7440-03-1 | Nb | 92.91 g/mol

Supplier:  Thermo Scientific Chemicals 04047822

 View more versions of this product

Catalog No. AA4047822


Only null left
Explore available promotions

Description

Description

Used in high temperature and super alloys of Zn and Ti

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Gray
3-1-7440
100 g
99.8% (Metals basis)
Nb
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
[Nb]
92.91
CHEBI:33344
Niobium
gray powder
Odorless
Powder
MFCD00011126
GUCVJGMIXFAOAE-UHFFFAOYSA-N
niobium
23936

Chemical Identifiers

3-1-7440
92.91
GUCVJGMIXFAOAE-UHFFFAOYSA-N
23936
niobium
Nb
MFCD00011126
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
CHEBI:33344
[Nb]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H228: Flammable solid.

DOTInformation : DOT Class: 4.1, Packaging Group: II

EINECSNumber : 231-113-5

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only