missing translation for 'onlineSavingsMsg'
Learn More

Niobium powder, APS 1-5 micron, 99.8% (metals basis)

Catalog No. AA4047809
Change view
Click to view available options
Quantity:
10 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4047809 10 g
AA4047822 100 g
AA4047830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4047809 Supplier Thermo Scientific Chemicals Supplier No. 04047809

CAS: 7440-03-1 | Nb | 92.91 g/mol

Used in high temperature and super alloys of Zn and Ti

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3-1-7440
Molecular Formula Nb
Molecular Weight (g/mol) 92.91
MDL Number MFCD00011126
InChI Key GUCVJGMIXFAOAE-UHFFFAOYSA-N
Synonym columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
PubChem CID 23936
ChEBI CHEBI:33344
IUPAC Name niobium
SMILES [Nb]

Specifications

Color Gray
Appearance gray powder
Density 8.57g/cm3 at 20°C
Quantity 10 g
Odor Odorless
Form Powder
Assay Percent Range 99.8% (Metals basis)
Chemical Name or Material Niobium
Hazard Category H228
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation DOT Class: 4.1, Packaging Group: II
EINECSNumber 231-113-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only