Learn More
Niobium powder, -60 mesh, 99.8% (metals basis excluding Ta), Ta <1%
CAS: 7440-03-1 | Nb | 92.91 g/mol
Supplier: Thermo Scientific Chemicals 01027622
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Silver to Gray | |
| 3-1-7440 | |
| 8.57g/cm3 at 20°C | |
| -60 Mesh | |
| Powder | |
| Nb | |
| columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio | |
| [Nb] | |
| 92.91 | |
| CHEBI:33344 |
| Niobium | |
| Silver gray powder | |
| 100 g | |
| Odorless | |
| 99.8% (Metals basis excluding Ta) | |
| MFCD00011126 | |
| GUCVJGMIXFAOAE-UHFFFAOYSA-N | |
| niobium | |
| 23936 |
Chemical Identifiers
| 3-1-7440 | |
| 92.91 | |
| GUCVJGMIXFAOAE-UHFFFAOYSA-N | |
| 23936 | |
| niobium |
| Nb | |
| MFCD00011126 | |
| columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio | |
| CHEBI:33344 | |
| [Nb] |
Safety and Handling
P210-P240-P241-P280-P370+P378q
H228
DOTInformation : Hazard Class: 4.1; Packaging Group: II
EINECSNumber : 231-113-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only