missing translation for 'onlineSavingsMsg'
Learn More

Niobium pieces, 99.8% (metals basis)

Catalog No. AA0024136
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA0024136 500 g
AA0024122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA0024136 Supplier Thermo Scientific Chemicals Supplier No. 00024136

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-03-1 | Nb | 92.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3-1-7440
Molecular Formula Nb
Molecular Weight (g/mol) 92.91
MDL Number MFCD00011126
InChI Key GUCVJGMIXFAOAE-UHFFFAOYSA-N
Synonym columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
PubChem CID 23936
ChEBI CHEBI:33344
IUPAC Name niobium
SMILES [Nb]

Specifications

Color Gray or Silver
Quantity 500 g
Odor Odorless
Form Pieces
Assay Percent Range 99.8% (metals basis)
Percent Purity 99.8%
Assay (metals basis)
Chemical Name or Material Niobium pieces
EINECSNumber 231-113-5
RTECSNumber QT9900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only