missing translation for 'onlineSavingsMsg'
Learn More

Niobium foil, 0.3mm (0.01in) thick, annealed, 99.8% (metals basis)

CAS: 7440-03-1 | Nb | 92.91 g/mol

Supplier:  Thermo Scientific Chemicals 040668GK

 View more versions of this product

Catalog No. AA40668GK


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Niobium foil
3-1-7440
Odorless
Foil
Nb
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
[Nb]
92.91
CHEBI:33344
0.3mm (0.01 in.) thick, Annealed
100 x 200 mm
≈12.9g/50x100mm
(metals basis)
MFCD00011126
GUCVJGMIXFAOAE-UHFFFAOYSA-N
niobium
23936
99.8%

Chemical Identifiers

3-1-7440
92.91
GUCVJGMIXFAOAE-UHFFFAOYSA-N
23936
niobium
Nb
MFCD00011126
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
CHEBI:33344
[Nb]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-113-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only