missing translation for 'onlineSavingsMsg'
Learn More

Niobium foil, 0.3mm (0.01in) thick, annealed, 99.8% (metals basis)

Catalog No. AA40668HQ
Change view
Click to view available options
Quantity:
50 x 100 mm
100 x 200 mm
200 x 500 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40668HQ 200 x 500 mm
AA40668FY 50 x 100 mm
AA40668GK 100 x 200 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA40668HQ Supplier Thermo Scientific Chemicals Supplier No. 040668HQ

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-03-1 | Nb | 92.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3-1-7440
Molecular Formula Nb
Molecular Weight (g/mol) 92.91
MDL Number MFCD00011126
InChI Key GUCVJGMIXFAOAE-UHFFFAOYSA-N
Synonym columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
PubChem CID 23936
ChEBI CHEBI:33344
IUPAC Name niobium
SMILES [Nb]

Specifications

Quantity 200 x 500 mm
Odor Odorless
Weight ≈12.9g/50x100mm
Form Foil
Percent Purity 99.8%
Assay (metals basis)
Chemical Name or Material Niobium foil, 0.3mm (0.01 in.) thick, Annealed
EINECSNumber 231-113-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only