missing translation for 'onlineSavingsMsg'
Learn More

Niobium foil, 0.25mm (0.01in) thick, 99.97% (metals basis excluding Ta), Ta ∣<0.06%

Catalog No. AA00238FF
Change view
Click to view available options
Quantity:
25 x 25 mm
25 x 100 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA00238FF 25 x 25 mm
AA00238FQ 25 x 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA00238FF Supplier Thermo Scientific Chemicals Supplier No. 000238FF

CAS: 7440-03-1 | Nb | 92.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3-1-7440
Molecular Formula Nb
Molecular Weight (g/mol) 92.91
MDL Number MFCD00011126
InChI Key GUCVJGMIXFAOAE-UHFFFAOYSA-N
Synonym columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
PubChem CID 23936
ChEBI CHEBI:33344
IUPAC Name niobium
SMILES [Nb]

Specifications

Color Gray or Silver
Quantity 25 x 25 mm
Odor Odorless
Weight ≈1.34g/25x25mm
Form Foil
Percent Purity 99.97%
Assay (metals basis excluding Ta), Ta ≤0.06%
Chemical Name or Material Niobium foil, 0.25mm (0.01 in.) thick
EINECSNumber 231-113-5
RTECSNumber QT9900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only