missing translation for 'onlineSavingsMsg'
Learn More

Niobium foil, 0.127mm (0.005in) thick, 99.97% (metals basis excluding Ta), Ta ∣<0.06%

CAS: 7440-03-1 | Nb | 92.91 g/mol

Supplier:  Thermo Scientific Chemicals 000236FQ

 View more versions of this product

Catalog No. AA00236FQ

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Gray or Silver
0.127mm (0.005 in.) thick
25 x 100 mm
≈0.68g/25x25mm
(metals basis excluding Ta), Ta ≤0.06%
MFCD00011126
GUCVJGMIXFAOAE-UHFFFAOYSA-N
niobium
23936
99.97%
Niobium foil
3-1-7440
Odorless
Foil
Nb
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
[Nb]
92.91
CHEBI:33344

Chemical Identifiers

3-1-7440
92.91
GUCVJGMIXFAOAE-UHFFFAOYSA-N
23936
niobium
Nb
MFCD00011126
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
CHEBI:33344
[Nb]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-113-5

RTECSNumber : QT9900000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only