missing translation for 'onlineSavingsMsg'
Learn More

Niobium foil, 0.05mm (0.002in) thick, annealed, 99.8% (metals basis)

Catalog No. p-7050912
Change view
Click to view available options
Quantity:
100 x 100 mm
100 x 500 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA39702GH 100 x 100 mm
AA39702HP 100 x 500 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA39702GH Supplier Thermo Scientific Chemicals Supplier No. 039702GH
Only null left

CAS: 7440-03-1 | Nb | 92.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3-1-7440
Molecular Formula Nb
Molecular Weight (g/mol) 92.91
MDL Number MFCD00011126
InChI Key GUCVJGMIXFAOAE-UHFFFAOYSA-N
Synonym columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
PubChem CID 23936
ChEBI CHEBI:33344
IUPAC Name niobium
SMILES [Nb]

Specifications

Quantity 100 x 100 mm
Odor Odorless
Weight ≈4.29g/100x100mm
Form Foil
Percent Purity 99.8%
Assay (metals basis)
Chemical Name or Material Niobium foil, 0.05mm (0.002 in.) thick, Annealed
EINECSNumber 231-113-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only