Please login to your online account to display your discounted pricing

Niobium AA Standard, 1mL = 1mg Nb (1,000ppm Nb)NH4NbF6 in Water / tr HF, Ricca Chemical


Quantity 100mL
Packaging Poly natural

Manufacturer: ricca chemical company  RIANB1KW100

 View more versions of this product

Catalog No. ANB1KW100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Color Clear colorless
Odor Odorless
Quantity 100mL
Chemical Name or Material Niobium AA Standard
Shelf Life 24 months
CAS Max % 0.1
CAS 7732-18-5
Concentration or Composition (by Analyte or Components) 1mL = 1mg Nb (1, 000ppm Nb)NH4NbF6 in Water / tr HF
CAS Min % 98.0
CAS Max % 100.0
CAS 12062-13-4
Packaging Poly natural
CAS Min % 0.25
CAS Max % 0.27
CAS 7664-39-3
CAS Min % 0.1

Specifications

Chemical Name or Material Niobium AA Standard
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 12062-13-4
CAS Min % 0.25
CAS Max % 0.27
CAS 7664-39-3
CAS Min % 0.1
CAS Max % 0.1
Accessories

24 months