missing translation for 'onlineSavingsMsg'
Learn More

Ninhydrin 98.0+%, TCI America™

Catalog No. p-7131157
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I001525G 25 g
I0015100G 100 g
I0015500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I001525G Supplier TCI America Supplier No. I001525G
Only null left

Chemical Identifiers

CAS 485-47-2
Molecular Formula C9H6O4
Molecular Weight (g/mol) 178.143
MDL Number MFCD00003791
InChI Key FEMOMIGRRWSMCU-UHFFFAOYSA-N
Synonym ninhydrin, ninhydrin hydrate, 1,2,3-indantrione monohydrate, ninhydrine, 2,2-dihydroxy-1h-indene-1,3 2h-dione, 2,2-dihydroxy-1,3-indandione, triketohydrindene hydrate, 1h-indene-1,3 2h-dione, 2,2-dihydroxy, 1,2,3-indantrione, 2-hydrate, 2,2-dihydroxyindane-1,3-dione
PubChem CID 10236
ChEBI CHEBI:86374
IUPAC Name 2,2-dihydroxyindene-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O

Specifications

Melting Point 250°C
Color White-Yellow
Quantity 25 g
Formula Weight 178.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ninhydrin
EINECSNumber (4)-0584
RTECSNumber NK5425000
TSCA Yes