missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ninhydrin, ozone friendly ready to use spray for TLC

Catalog No. AC327022400
Change view
Click to view available options
Quantity:
240 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC327022400 240 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC327022400 Supplier Thermo Scientific Chemicals Supplier No. 327022400
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 485-47-2 | C9H6O4 | 178.143 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Ninhydrin
Boiling Point >34.0°C
CAS 485-47-2, 106-97-8, 74-98-6, 71-36-3
Packaging Spray can
Molecular Formula C9H6O4
MDL Number MFCD00003791
Density 0.81
Merck Index 15, 6640
Quantity 240 mL
InChI Key FEMOMIGRRWSMCU-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O
IUPAC Name 2,2-dihydroxyindene-1,3-dione
Molecular Weight (g/mol) 178.143
PubChem CID 10236
ChEBI CHEBI:86374
Show More Show Less

RUO – Research Use Only