missing translation for 'onlineSavingsMsg'
Learn More

Ninhydrin, EMD-Millipore

Catalog No. M1067050100
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1067050100 100 mL Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1067050100 Supplier Technidata Supplier No. 1.06705.0100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ready-to-use spray solutions for thin-layer chromatography

Chemical Identifiers

CAS 485-47-2
Molecular Formula C9H6O4
Molecular Weight (g/mol) 178.143
InChI Key FEMOMIGRRWSMCU-UHFFFAOYSA-N
Synonym ninhydrin, ninhydrin hydrate, 1,2,3-indantrione monohydrate, ninhydrine, 2,2-dihydroxy-1h-indene-1,3 2h-dione, 2,2-dihydroxy-1,3-indandione, triketohydrindene hydrate, 1h-indene-1,3 2h-dione, 2,2-dihydroxy, 1,2,3-indantrione, 2-hydrate, 2,2-dihydroxyindane-1,3-dione
PubChem CID 10236
ChEBI CHEBI:86374
IUPAC Name 2,2-dihydroxyindene-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O

Specifications

Explosion Limit 2 to 13.4 %(V) (2-Propanol)
Ignition Point 425°C (2-Propanol)
Packaging Glass
Quantity 100 mL
Vapor Pressure 42.4 hPa (20°C)
Chemical Name or Material Ninhydrin
Recommended Storage Store at 15° to 25°C