missing translation for 'onlineSavingsMsg'
Learn More

Ninhydrin (contains Acetic Acid) Ethanol Solution [for TLC Stain], TCI America™

Catalog No. N0719100ML
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0719100ML 100 mL
N0719500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0719100ML Supplier TCI America Supplier No. N0719100ML

Chemical Identifiers

CAS 485-47-2
Molecular Formula C9H6O4
Molecular Weight (g/mol) 178.143
MDL Number MFCD00003791
InChI Key FEMOMIGRRWSMCU-UHFFFAOYSA-N
Synonym ninhydrin, ninhydrin hydrate, 1,2,3-indantrione monohydrate, ninhydrine, 2,2-dihydroxy-1h-indene-1,3 2h-dione, 2,2-dihydroxy-1,3-indandione, triketohydrindene hydrate, 1h-indene-1,3 2h-dione, 2,2-dihydroxy, 1,2,3-indantrione, 2-hydrate, 2,2-dihydroxyindane-1,3-dione
PubChem CID 10236
ChEBI CHEBI:86374
IUPAC Name 2,2-dihydroxyindene-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O

Specifications

Color Yellow
Quantity 100 mL
UN Number 1170
Formula Weight 178.14
Physical Form Liquid
Chemical Name or Material Ninhydrin (contains Acetic Acid) Ethanol Solution [for TLC Stain]
EINECSNumber (4)-0584
RTECSNumber NK5425000
TSCA Yes