Learn More
Ninhydrin, 99%
CAS: 485-47-2 | C9H6O4 | 178.143 g/mol
Supplier: Thermo Scientific Chemicals A1040918
Description
Ninhydrin is used for the determination of amino groups in amino acids, peptides and proteins. It finds application in forensic science for the analysis of latent fingerprints on porous surfaces such as paper. It is also used in thin layer chromatography (TLC) to monitor the progress of chemical reactions. In Kaiser test, it plays an important role to monitor the deprotection in solid phase peptide synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNinhydrin is used for the determination of amino groups in amino acids, peptides and proteins. It finds application in forensic science for the analysis of latent fingerprints on porous surfaces such as paper. It is also used in thin layer chromatography (TLC) to monitor the progress of chemical reactions. In Kaiser test, it plays an important role to monitor the deprotection in solid phase peptide synthesis.
Solubility
Soluble in water, acetone, ethanol, and methanol. Slightly soluble in chloroform and ether.
Notes
Light Sensitive. Incompatible with strong oxidizing agents, amines, alkalis and strong bases.
Specifications
| Ninhydrin | |
| 251°C (decomposition) | |
| C9H6O4 | |
| 50 g | |
| Light sensitive | |
| ninhydrin, ninhydrin hydrate, 1,2,3-indantrione monohydrate, ninhydrine, 2,2-dihydroxy-1h-indene-1,3 2h-dione, 2,2-dihydroxy-1,3-indandione, triketohydrindene hydrate, 1h-indene-1,3 2h-dione, 2,2-dihydroxy, 1,2,3-indantrione, 2-hydrate, 2,2-dihydroxyindane-1,3-dione | |
| FEMOMIGRRWSMCU-UHFFFAOYSA-N | |
| 2,2-dihydroxyindene-1,3-dione | |
| 10236 | |
| 178.14 | |
| Powder |
| 485-47-2 | |
| 99% | |
| MFCD00003791 | |
| 1910963 | |
| 14,6554 | |
| Soluble in water,acetone,ethanol,and methanol. Slightly soluble in chloroform and ether. | |
| C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O | |
| 178.143 | |
| CHEBI:86374 | |
| 99% |
Chemical Identifiers
| 485-47-2 | |
| 178.143 | |
| FEMOMIGRRWSMCU-UHFFFAOYSA-N | |
| 10236 | |
| 2,2-dihydroxyindene-1,3-dione |
| C9H6O4 | |
| MFCD00003791 | |
| ninhydrin, ninhydrin hydrate, 1,2,3-indantrione monohydrate, ninhydrine, 2,2-dihydroxy-1h-indene-1,3 2h-dione, 2,2-dihydroxy-1,3-indandione, triketohydrindene hydrate, 1h-indene-1,3 2h-dione, 2,2-dihydroxy, 1,2,3-indantrione, 2-hydrate, 2,2-dihydroxyindane-1,3-dione | |
| CHEBI:86374 | |
| C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 207-618-1
RTECSNumber : NK5425000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only