missing translation for 'onlineSavingsMsg'
Learn More

Ninhydrin, 99%

CAS: 485-47-2 | C9H6O4 | 178.143 g/mol

Supplier:  Thermo Scientific Chemicals A1040914

 View more versions of this product

Catalog No. AAA1040914

Only null left
Explore available promotions

Description

Description

Ninhydrin is used for the determination of amino groups in amino acids, peptides and proteins. It finds application in forensic science for the analysis of latent fingerprints on porous surfaces such as paper. It is also used in thin layer chromatography (TLC) to monitor the progress of chemical reactions. In Kaiser test, it plays an important role to monitor the deprotection in solid phase peptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ninhydrin is used for the determination of amino groups in amino acids, peptides and proteins. It finds application in forensic science for the analysis of latent fingerprints on porous surfaces such as paper. It is also used in thin layer chromatography (TLC) to monitor the progress of chemical reactions. In Kaiser test, it plays an important role to monitor the deprotection in solid phase peptide synthesis.

Solubility
Soluble in water, acetone, ethanol, and methanol. Slightly soluble in chloroform and ether.

Notes
Light Sensitive. Incompatible with strong oxidizing agents, amines, alkalis and strong bases.
Specifications

Specifications

Ninhydrin
251°C (decomposition)
C9H6O4
25 g
Light sensitive
ninhydrin, ninhydrin hydrate, 1,2,3-indantrione monohydrate, ninhydrine, 2,2-dihydroxy-1h-indene-1,3 2h-dione, 2,2-dihydroxy-1,3-indandione, triketohydrindene hydrate, 1h-indene-1,3 2h-dione, 2,2-dihydroxy, 1,2,3-indantrione, 2-hydrate, 2,2-dihydroxyindane-1,3-dione
FEMOMIGRRWSMCU-UHFFFAOYSA-N
2,2-dihydroxyindene-1,3-dione
10236
178.14
Powder
485-47-2
99%
MFCD00003791
1910963
14,6554
Soluble in water,acetone,ethanol,and methanol. Slightly soluble in chloroform and ether.
C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O
178.143
CHEBI:86374
99%

Chemical Identifiers

485-47-2
178.143
FEMOMIGRRWSMCU-UHFFFAOYSA-N
10236
2,2-dihydroxyindene-1,3-dione
C9H6O4
MFCD00003791
ninhydrin, ninhydrin hydrate, 1,2,3-indantrione monohydrate, ninhydrine, 2,2-dihydroxy-1h-indene-1,3 2h-dione, 2,2-dihydroxy-1,3-indandione, triketohydrindene hydrate, 1h-indene-1,3 2h-dione, 2,2-dihydroxy, 1,2,3-indantrione, 2-hydrate, 2,2-dihydroxyindane-1,3-dione
CHEBI:86374
C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 207-618-1

RTECSNumber : NK5425000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only