missing translation for 'onlineSavingsMsg'
Learn More

Ninhydrin, (1,2,3-Indantrione Monohydrate), Reagents

Catalog No. 01337155
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337155 25 g
01337154 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337155 Supplier Labchem Inc Supplier No. C225700025C1
Only null left

Ninhydrin, (1,2,3-Indantrione Monohydrate), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 485-47-2
Molecular Formula C9H6O4
Molecular Weight (g/mol) 178.14
InChI Key FEMOMIGRRWSMCU-UHFFFAOYSA-N
Synonym 1,2,3-Triketohydrindene Monohydrate, Ninhydrine, 1,2,3-Indantrione Monohydrate
IUPAC Name 2,2-dihydroxy-2,3-dihydro-1H-indene-1,3-dione
SMILES OC1(O)C(=O)C2=CC=CC=C2C1=O

Specifications

Color White to brownish-White
Packaging Glass Bottle
Quantity 25 g
Specific Gravity 0.862
Grade Reagent
Physical Form Solid
Chemical Name or Material Ninhydrin, (1,2,3-Indantrione Monohydrate)