Learn More
Nine-Liter Wet Spirometer
Catalog No. p-2370095
Change view
Click to view available options
Product Type:
Mouthpiece
Quantity:
100 Pack
Catalog No. Product Type Quantity
S17046A Mouthpiece 100 Pack
1 options
This item is intended for educational purposes only.
Catalog No. S17046A Supplier Customer Channels Group (Ccg) Supplier No. 7085 301 SU
Only null left
This item is intended for educational purposes only.
Specially designed to meet the needs of today's physiology teachers.
This academic-quality wet spirometer combines reliability and accuracy with an increased volume to accommodate students with larger lung capacities.
  • Large, easy-to-read scale graduations
  • Includes disposable mouthpieces and a one-way plastic valve for sanitary classroom use
  • Made of lightweight, yet durable, high-density polyethylene (HDPE) plastic and aluminum
Determine Five Different Respiratory Volumes:
  • Tidal Volume (TV)
  • Inspiratory Capacity (IC)
  • Inspiratory Reserve Volume (IRV)
  • Expiratory Reserve Volume (ERV)
  • Vital Capacity (VC)
Specifications
Quantity 100 Pack
Disposable Single-use
For Use With (Application) For measuring the volume of air inspired and expired by the lungs
Product Type Mouthpiece
For Use With (Equipment) Student wet spirometer