missing translation for 'onlineSavingsMsg'
Learn More

Nimodipine, 98+%

Catalog No. p-7042079
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61287ME 500 mg
AAJ6128703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61287ME Supplier Thermo Scientific Chemicals Supplier No. J61287ME
Only null left

Potent L-type calcium channel antagonist | CAS: 66085-59-4 | C21H26N2O7 | 418.45 g/mol

Potent L-type calcium channel antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 66085-59-4
Molecular Formula C21H26N2O7
Molecular Weight (g/mol) 418.45
MDL Number MFCD00153848
InChI Key UIAGMCDKSXEBJQ-UHFFFAOYNA-N
Synonym nimodipine, nimotop, periplum, nimodipino, nimodipinum, nymalize, admon, nimodipinum inn-latin, nimodipino inn-spanish, isopropyl 2-methoxyethyl 1,4-dihydro-2,6-dimethyl-4-m-nitrophenyl-3,5-pyridinedicarboxylate
PubChem CID 4497
ChEBI CHEBI:7575
IUPAC Name 3-(2-methoxyethyl) 5-propan-2-yl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
SMILES COCCOC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OC(C)C

Specifications

Color Yellow
Quantity 500 mg
Merck Index 14,6551
Formula Weight 418.44
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Nimodipine
Hazard Category H373
Hazard Statement GHS H Statement
H312-H332
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P260-P314-P501c
EINECSNumber 266-127-0
RTECSNumber US7975500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only