missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Nile Blue A
Description
Used in the preparation of an amperometric glucose sensor. Also can be employed in Lillie and Fullmer method for staining and discriminating between melanins and lipofuscins in paraffin sections of animal tissue; Lillie and Fuller method for fatty acids/lipids using frozen sections of fatty liver and avocado.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used in the preparation of an amperometric glucose sensor. Also can be employed in Lillie and Fullmer method for staining and discriminating between melanins and lipofuscins in paraffin sections of animal tissue; Lillie and Fuller method for fatty acids/lipids using frozen sections of fatty liver and avocado.
Solubility
Soluble in ethanol, and water (50 g/L) at 25°C.
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
Used in the preparation of an amperometric glucose sensor. Also can be employed in Lillie and Fullmer method for staining and discriminating between melanins and lipofuscins in paraffin sections of animal tissue; Lillie and Fuller method for fatty acids/lipids using frozen sections of fatty liver and avocado.
Solubility
Soluble in ethanol, and water (50 g/L) at 25°C.
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
Specifications
Specifications
| Melting Point | >300°C (decomposition) |
| Synonym | C.I. 51180, Basic Blue 12 |
| Solubility Information | Soluble in ethanol,and water (50g/L) at 25°C. |
| Formula Weight | 732.84 |
| Quantity | 5 g |
| Chemical Name or Material | Nile Blue A |
Safety and Handling
| EINECSNumber | 222-832-5 |
| RTECSNumber | DJ1931000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only