missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nigrosine, pure, water soluble, high purity biological stain

CAS: 8005-03-6 | C22H14N6Na2O9S2 | 616.49 g/mol

Supplier:  Thermo Scientific Chemicals 189481000

 View more versions of this product

Catalog No. AC189481000


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Nigrosine
8005-03-6
MFCD00044681
Acid Black 2, C.I. 50420
HKBVRFLHNUEVRO-HMDKGIQMSA-L
C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
616.49
Pure
Complies
100 g
Water soluble, Pure
C22H14N6Na2O9S2
12, 130
Solubility in water: soluble.
Authentic
disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
44134531
Glass bottle
Black
Crystalline Powder

Chemical Identifiers

8005-03-6
616.49
HKBVRFLHNUEVRO-HMDKGIQMSA-L
44134531
C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
C22H14N6Na2O9S2
MFCD00044681
Acid Black 2, C.I. 50420
disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only