missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nigrosine, pure, water soluble, high purity biological stain

Catalog No. AC189480250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC189480250 25 g Glass bottle
AC189481000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC189480250 Supplier Thermo Scientific Chemicals Supplier No. 189480250

CAS: 8005-03-6 | C22H14N6Na2O9S2 | 616.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 8005-03-6
Molecular Formula C22H14N6Na2O9S2
Molecular Weight (g/mol) 616.49
MDL Number MFCD00044681
InChI Key HKBVRFLHNUEVRO-HMDKGIQMSA-L
Synonym Acid Black 2, C.I. 50420
PubChem CID 44134531
IUPAC Name disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Beilstein 12, 130
Solubility Information Solubility in water: soluble.
Infrared Spectrum Authentic
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Black
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Nigrosine, Water soluble, Pure

RUO – Research Use Only