missing translation for 'onlineSavingsMsg'
Learn More

Nigrosin water soluble

Catalog No. AAA1814714
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1814714 25 g
AAA1814722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1814714 Supplier Thermo Scientific Chemicals Supplier No. A1814714

CAS: 8005-03-6 | C22H14N6Na2O9S2 | 616.49 g/mol

It is employed as a common biological stain for tissues and cells. Used to make inks and shoe-polish and for dyeing leather, wood, textiles, and plastics. The crude mixture of nigrosine hydrochlorides (Phenol Black L) is used to dye phenolic resins. Bases are used to make printing colors and to dye plastics. It finds its use in Dorner?s spore stain, as a negative stain in place of India ink for spirochetes, bacteria, protozoa and fungi. It is also used in compounding resins include phenolic resins, styrenics, polyamides and urea resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a common biological stain for tissues and cells. Used to make inks and shoe-polish and for dyeing leather, wood, textiles, and plastics. The crude mixture of nigrosine hydrochlorides (Phenol Black L) is used to dye phenolic resins. Bases are used to make printing colors and to dye plastics. It finds its use in Dorner′s spore stain, as a negative stain in place of India ink for spirochetes, bacteria, protozoa and fungi. It is also used in compounding resins include phenolic resins, styrenics, polyamides and urea resins.

Solubility
Soluble in water, ethanol, oil, nigrosines and fat.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 8005-03-6
Molecular Formula C22H14N6Na2O9S2
Molecular Weight (g/mol) 616.49
MDL Number MFCD00044681
InChI Key HKBVRFLHNUEVRO-HMDKGIQMSA-L
PubChem CID 44134531
IUPAC Name disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Solubility Information Soluble in water,ethanol,oil,nigrosines and fat.
Quantity 25 g
Chemical Name or Material Nigrosin, water soluble
RTECSNumber GC4762250
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only