missing translation for 'onlineSavingsMsg'
Learn More

Nigrosin, alcohol soluble

CAS: 11099-03-9 | C8H19ClN2O | 194.703 g/mol

Supplier:  Thermo Scientific Chemicals J61186A1

 View more versions of this product

Catalog No. AAJ61186A1

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Nigrosin
C8H19ClN2O
Solvent Black 5; C.I. 50415
GBPWTMZRCAESKB-UHFFFAOYSA-N
2-amino-N,N-dipropylacetamide;hydrochloride
19828004
1 kg
11099-03-9
MFCD00149005
Soluble in alcohol
CCCN(CCC)C(=O)CN.Cl
194.703
Black
Powder

Chemical Identifiers

11099-03-9
194.703
GBPWTMZRCAESKB-UHFFFAOYSA-N
19828004
CCCN(CCC)C(=O)CN.Cl
C8H19ClN2O
MFCD00149005
Solvent Black 5; C.I. 50415
2-amino-N,N-dipropylacetamide;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : GE5800000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only