missing translation for 'onlineSavingsMsg'
Learn More

Nigrosin, alcohol soluble

Catalog No. AAJ6118614
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6118614 25 g
AAJ6118622 100 g
AAJ61186A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6118614 Supplier Thermo Scientific Chemicals Supplier No. J6118614

CAS: 11099-03-9 | C8H19ClN2O | 194.703 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 11099-03-9
Molecular Formula C8H19ClN2O
Molecular Weight (g/mol) 194.703
MDL Number MFCD00149005
InChI Key GBPWTMZRCAESKB-UHFFFAOYSA-N
Synonym Solvent Black 5; C.I. 50415
PubChem CID 19828004
IUPAC Name 2-amino-N,N-dipropylacetamide;hydrochloride
SMILES CCCN(CCC)C(=O)CN.Cl

Specifications

Solubility Information Soluble in alcohol
Color Black
Quantity 25 g
Physical Form Powder
Chemical Name or Material Nigrosin
RTECSNumber GE5800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only