Learn More
Nicotinic acid, specified according to the requirements of Ph. Eur, Thermo Scientific Chemicals
$462.93 - $462.93
Chemical Identifiers
| CAS | 59-67-6 |
|---|---|
| Molecular Formula | C6H5NO2 |
| Molecular Weight (g/mol) | 123.11 |
| MDL Number | MFCD00006391 |
| InChI Key | PVNIIMVLHYAWGP-UHFFFAOYSA-N |
| Synonym | nicotinic acid, niacin, 3-pyridinecarboxylic acid, 3-carboxypyridine, wampocap, acidum nicotinicum, apelagrin, pellagrin, akotin, daskil |
| PubChem CID | 938 |
| ChEBI | CHEBI:15940 |
| IUPAC Name | pyridine-3-carboxylic acid |
| SMILES | OC(=O)C1=CC=CN=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC380325000
|
Thermo Scientific Chemicals
380325000 |
500 g | Plastic bottle |
Each for $462.93
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 59-67-6 | |
| 123.11 | |
| PVNIIMVLHYAWGP-UHFFFAOYSA-N | |
| 938 | |
| pyridine-3-carboxylic acid |
| C6H5NO2 | |
| MFCD00006391 | |
| nicotinic acid, niacin, 3-pyridinecarboxylic acid, 3-carboxypyridine, wampocap, acidum nicotinicum, apelagrin, pellagrin, akotin, daskil | |
| CHEBI:15940 | |
| OC(=O)C1=CC=CN=C1 |
Specifications
| 59-67-6 | |
| Plastic bottle | |
| MFCD00006391 | |
| 15,661 | |
| (in water) Sparingly soluble | |
| OC(=O)C1=CC=CN=C1 | |
| 123.11 | |
| CHEBI:15940 | |
| Passes test | |
| Nicotinic acid |
| 236.0°C to 239.0°C | |
| C6H5NO2 | |
| 500 g | |
| nicotinic acid, niacin, 3-pyridinecarboxylic acid, 3-carboxypyridine, wampocap, acidum nicotinicum, apelagrin, pellagrin, akotin, daskil | |
| PVNIIMVLHYAWGP-UHFFFAOYSA-N | |
| pyridine-3-carboxylic acid | |
| 938 | |
| 123.11 | |
| Specified according to the requirements of Ph. Eur |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 200-441-
RUO – Research Use Only