missing translation for 'onlineSavingsMsg'
Learn More

Nicotinic Acid N-Oxide 98.0+%, TCI America™

Catalog No. p-7133838
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N049725G 25 g
N0497500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N049725G Supplier TCI America Supplier No. N049725G
Only null left

Chemical Identifiers

CAS 2398-81-4
Molecular Formula C6H6NO3
Molecular Weight (g/mol) 140.12
MDL Number MFCD00006201
InChI Key GDILJONVBZVAFH-UHFFFAOYNA-N
Synonym nicotinic acid n-oxide, oxiniacic acid, nicotinic acid 1-oxide, 3-pyridinecarboxylic acid, 1-oxide, nicotinic acid oxide, n-hydroxynicotinic acid, 3-carboxypyridin-1-ium-1-olate, 3-carboxypyridine n-oxide, 3-carboxypyridine 1-oxide, 3-pyridinecarboxylic acid oxide
PubChem CID 16976
IUPAC Name 5-carboxy-1,2-dihydro-1λ²-pyridine-1,1-diium-2-id-1-olate
SMILES OC(=O)C1=C[NH++]([O-])[CH-]C=C1

Specifications

Melting Point 255°C
Quantity 25 g
Formula Weight 139.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nicotinic Acid N-Oxide