missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinic Acid N-Oxide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 255°C |
| Quantity | 25 g |
| Formula Weight | 139.11 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Nicotinic Acid N-Oxide |
Safety and Handling
| TSCA | Yes |