missing translation for 'onlineSavingsMsg'
Learn More

Nicotinic Acid 99.5+%, TCI America™

Catalog No. N11035G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N11035G 5 g
N11031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N11035G Supplier TCI America Supplier No. N11035G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 59-67-6 | C6H5NO2 | 123.11 g/mol

Chemical Identifiers

CAS 59-67-6
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.11
MDL Number MFCD00006391
InChI Key PVNIIMVLHYAWGP-UHFFFAOYSA-N
Synonym nicotinic acid, niacin, 3-pyridinecarboxylic acid, 3-carboxypyridine, wampocap, acidum nicotinicum, apelagrin, pellagrin, akotin, daskil
PubChem CID 938
ChEBI CHEBI:15940
IUPAC Name pyridine-3-carboxylic acid
SMILES OC(=O)C1=CC=CN=C1

Specifications

Melting Point 238°C
Color White
pH 2.7 (saturated H2O soln.)
Quantity 5 g
Formula Weight 123.11
Percent Purity ≥99.5% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nicotinic Acid [Matrix for MALDI-TOF/MS]
EINECSNumber (5)-0731
RTECSNumber QT0525000
TSCA Yes