missing translation for 'onlineSavingsMsg'
Learn More

Nicotinic Acid Hydrazide 98.0+%, TCI America™

Catalog No. N008725G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N008725G 25 g
N0087100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N008725G Supplier TCI America Supplier No. N008725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 553-53-7
Molecular Formula C6H7N3O
Molecular Weight (g/mol) 137.14
MDL Number MFCD00006383
InChI Key KFUSANSHCADHNJ-UHFFFAOYSA-N
Synonym nicotinohydrazide, nicotinic acid hydrazide, nicotinic hydrazide, niazid, nicotinylhydrazide, nicotinoyl hydrazine, 3-pyridoyl hydrazine, 3-pyridylcarbonylhydrazine, nicotinoyl hydrazide, hydrazine, nicotinoyl
PubChem CID 11112
IUPAC Name pyridine-3-carbohydrazide
SMILES NNC(=O)C1=CC=CN=C1

Specifications

Melting Point 162°C
Color White
Quantity 25 g
Formula Weight 137.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nicotinic Acid Hydrazide
RTECSNumber QT1750000
TSCA No