Learn More
Nicotinic acid, 99%
CAS: 59-67-6 | C6H5NO2 | 123.11 g/mol
Supplier: Thermo Scientific Chemicals A126830B
Description
Vitamin of the B complex with hypolipidemic properties occurring in various animal and plant tissues. Required by the body for the formation of coenzymes NAD and NADP.Nicotinic acid is used as an antidyslipidemic agent. It is also used to improve all lipoproteins well above the vitamin requirement and lowers the total cholesterol, bad LDL-cholesterol, and triglyceride levels. Further, it is used as a food additive in food industries, cosmetics and pharmaceuticals. In addition to this, it is used in medication, nutrient, enriched flours and in electroplating baths.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsVitamin of the B complex with hypolipidemic properties occurring in various animal and plant tissues. Required by the body for the formation of coenzymes NAD and NADP.Nicotinic acid is used as an antidyslipidemic agent. It is also used to improve all lipoproteins well above the vitamin requirement and lowers the total cholesterol, ″bad″ LDL-cholesterol, and triglyceride levels. Further, it is used as a food additive in food industries, cosmetics and pharmaceuticals. In addition to this, it is used in medication, nutrient, enriched flours and in electroplating baths.
Solubility
Soluble in water. Slightly soluble in ethanol and ethyl ether. Insoluble in most lipid solvents.
Notes
Light sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
| Nicotinic acid | |
| 236°C to 239°C | |
| 2.7 | |
| 99% | |
| C6H5NO2 | |
| 1000 g | |
| 14,6525 | |
| Soluble in water. Slightly soluble in ethanol and ethyl ether. Insoluble in most lipid solvents. | |
| OC(=O)C1=CC=CN=C1 | |
| 123.11 | |
| CHEBI:15940 | |
| 99% |
| 59-67-6 | |
| 1.4 | |
| 293°C (559°F) | |
| Odorless | |
| MFCD00006391 | |
| 109591 | |
| nicotinic acid, niacin, 3-pyridinecarboxylic acid, 3-carboxypyridine, wampocap, acidum nicotinicum, apelagrin, pellagrin, akotin, daskil | |
| PVNIIMVLHYAWGP-UHFFFAOYSA-N | |
| pyridine-3-carboxylic acid | |
| 938 | |
| 123.11 |
Chemical Identifiers
| 59-67-6 | |
| 123.11 | |
| PVNIIMVLHYAWGP-UHFFFAOYSA-N | |
| 938 | |
| pyridine-3-carboxylic acid |
| C6H5NO2 | |
| MFCD00006391 | |
| nicotinic acid, niacin, 3-pyridinecarboxylic acid, 3-carboxypyridine, wampocap, acidum nicotinicum, apelagrin, pellagrin, akotin, daskil | |
| CHEBI:15940 | |
| OC(=O)C1=CC=CN=C1 |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P264b-P280i-P305+P351+P338
H319
EINECSNumber : 200-441-0
RTECSNumber : QT0525000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only