missing translation for 'onlineSavingsMsg'
Learn More

Nicotinic Acid, ≥99%, MP Biomedicals™

$51.74 - $51.74

Chemical Identifiers

CAS 59-67-6
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.11
MDL Number MFCD00006391
InChI Key PVNIIMVLHYAWGP-UHFFFAOYSA-N
Synonym nicotinic acid, niacin, 3-pyridinecarboxylic acid, 3-carboxypyridine, wampocap, acidum nicotinicum, apelagrin, pellagrin, akotin, daskil
PubChem CID 938
ChEBI CHEBI:15940
IUPAC Name pyridine-3-carboxylic acid
SMILES OC(=O)C1=CC=CN=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN-10244680
View Documents
MP Biomedicals Inc
0210244680
100 g
Each for $51.74
Only null left
 
Description

Description

Specifications

Chemical Identifiers

59-67-6
123.11
PVNIIMVLHYAWGP-UHFFFAOYSA-N
938
pyridine-3-carboxylic acid
C6H5NO2
MFCD00006391
nicotinic acid, niacin, 3-pyridinecarboxylic acid, 3-carboxypyridine, wampocap, acidum nicotinicum, apelagrin, pellagrin, akotin, daskil
CHEBI:15940
OC(=O)C1=CC=CN=C1

Specifications

59-67-6
≥99%
MFCD00006391
nicotinic acid, niacin, 3-pyridinecarboxylic acid, 3-carboxypyridine, wampocap, acidum nicotinicum, apelagrin, pellagrin, akotin, daskil
OC(=O)C1=CC=CN=C1
123.11
CHEBI:15940
5.7 x 10-6mm Hg at 25°C (estimated)(Lit.)
Nicotinic Acid
White
C6H5NO2
100 g
PVNIIMVLHYAWGP-UHFFFAOYSA-N
pyridine-3-carboxylic acid
938
123.1
Solid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-441-0

Recommended Storage : Store at room temperature (15° to 30°C)

SDS
Documents

Documents