missing translation for 'onlineSavingsMsg'
Learn More

Nicotinic Acid, ≥99%, MP Biomedicals™

Catalog No. ICN10244680
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10244680 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10244680 Supplier MP Biomedicals Inc Supplier No. 0210244680

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 59-67-6
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.11
MDL Number MFCD00006391
InChI Key PVNIIMVLHYAWGP-UHFFFAOYSA-N
Synonym nicotinic acid, niacin, 3-pyridinecarboxylic acid, 3-carboxypyridine, wampocap, acidum nicotinicum, apelagrin, pellagrin, akotin, daskil
PubChem CID 938
ChEBI CHEBI:15940
IUPAC Name pyridine-3-carboxylic acid
SMILES OC(=O)C1=CC=CN=C1

Specifications

Color White
Assay Percent Range ≥99%
Quantity 100 g
Vapor Pressure 5.7 x 10-6mm Hg at 25°C (estimated)(Lit.)
Formula Weight 123.1
Physical Form Solid
Chemical Name or Material Nicotinic Acid
EINECSNumber 200-441-0
Recommended Storage Store at room temperature (15° to 30°C)