missing translation for 'onlineSavingsMsg'
Learn More

Nicotinic acid, 99.5%

Catalog No. AC128290100
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
10 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC128290100 10 kg Plastic drum
AC128290050 5 g Glass Bottle
AC128291000 100 g Plastic bottle
AC128295000 500 g Plastic bottle
AC128290025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC128290100 Supplier Thermo Scientific Chemicals Supplier No. 128290100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 59-67-6 | C6H5NO2 | 123.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59-67-6
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.11
MDL Number MFCD00006391
InChI Key PVNIIMVLHYAWGP-UHFFFAOYSA-N
Synonym nicotinic acid, niacin, 3-pyridinecarboxylic acid, 3-carboxypyridine, wampocap, acidum nicotinicum, apelagrin, pellagrin, akotin, daskil
PubChem CID 938
ChEBI CHEBI:15940
IUPAC Name pyridine-3-carboxylic acid
SMILES OC(=O)C1=CC=CN=C1

Specifications

CAS Min % 99.4
CAS Max % 100.0
Melting Point 236.0°C to 239.0°C
Color White
Loss on Drying 1% max. (105°C, 1 hr.)
Infrared Spectrum Authentic
Assay Percent Range 99.5%
Packaging Plastic drum
Quantity 10 kg
Beilstein 22,38
Merck Index 15,661
Solubility Information Solubility in water: 17g/L (20°C). Other solubilities: slightly soluble in ethanol (10g/L at 20°C),eth,er,soluble in propylene glycol,in alkali hydroxides,and carbonates,freely soluble in boiling water or boiling ethanol
Formula Weight 123.11
Percent Purity 99.5%
Physical Form Powder
Chemical Name or Material Nicotinic acid, 99.5%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 200-441-0
RTECSNumber QT0525000
TSCA TSCA

RUO – Research Use Only