missing translation for 'onlineSavingsMsg'
Learn More

Nicotine 95.0+%, TCI America™

Catalog No. p-7133604
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N007925ML 25 mL
N0079100ML 100 mL
N0079500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Catalog No. N007925ML missing translation for 'mfr' TCI America missing translation for 'supplierNo' N007925ML
Only null left

Chemical Identifiers

CAS 54-11-5
Molecular Formula C10H14N2
Molecular Weight (g/mol) 162.236
MDL Number MFCD00006369
InChI Key SNICXCGAKADSCV-JTQLQIEISA-N
Synonym nicotine, l-nicotine, --nicotine, s-nicotine, habitrol, s-3-1-methylpyrrolidin-2-yl pyridine, nicoderm, nicotrol, s---nicotine, nicoderm cq
PubChem CID 89594
ChEBI CHEBI:17688
IUPAC Name 3-[(2S)-1-methylpyrrolidin-2-yl]pyridine
SMILES CN1CCCC1C2=CN=CC=C2

Specifications

Melting Point -80°C
pH 10.2 (8.1g/L H2O soln. 20°C)
Boiling Point 91°C
Quantity 25 mL
UN Number 1654
Formula Weight 162.24
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Nicotine
EINECSNumber (9)-0990
RTECSNumber QS5250000
TSCA Yes