missing translation for 'onlineSavingsMsg'
Learn More

Nicotine 95.0+%, TCI America™

Catalog No. N0079500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0079500ML 500 mL
N007925ML 25 mL
N0079100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N0079500ML Supplier TCI America Supplier No. N0079500ML

Chemical Identifiers

CAS 54-11-5
Molecular Formula C10H14N2
Molecular Weight (g/mol) 162.236
MDL Number MFCD00006369
InChI Key SNICXCGAKADSCV-JTQLQIEISA-N
Synonym nicotine, l-nicotine, --nicotine, s-nicotine, habitrol, s-3-1-methylpyrrolidin-2-yl pyridine, nicoderm, nicotrol, s---nicotine, nicoderm cq
PubChem CID 89594
ChEBI CHEBI:17688
IUPAC Name 3-[(2S)-1-methylpyrrolidin-2-yl]pyridine
SMILES CN1CCCC1C2=CN=CC=C2

Specifications

Melting Point -80°C
pH 10.2 (8.1g/L H2O soln. 20°C)
Boiling Point 91°C
Quantity 500 mL
UN Number 1654
Formula Weight 162.24
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Nicotine
EINECSNumber (9)-0990
RTECSNumber QS5250000
TSCA Yes