missing translation for 'onlineSavingsMsg'
Learn More

Nicotine ditartrate dihydrate, 98%

CAS: 6019-06-3 | C18H30N2O14 | 498.44 g/mol

Supplier:  Thermo Scientific Chemicals 415660100

 View more versions of this product

Catalog No. AC415660100


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Nicotine ditartrate dihydrate
6-3-6019
100.0
White to Yellow
98%
C18H30N2O14
10 g
Solubility in water: 50g/L (20°C)
O.O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.CN1CCC[C@H]1C1=CN=CC=C1
498.44
98%
98%
97.5
97.0°C to 100.0°C
Authentic
Glass bottle
MFCD00150899
(-)-Nicotine di-(+)-hydrogen tartrate dihydrate
LDMPZNTVIGIREC-HYRPJEHINA-N
N-{3-[(2Z)-5-chloro-2,3-dihydro-1,3-benzoxazol-2-ylidene]-4-oxocyclohexa-1,5-dien-1-yl}-2-methylpropanamide
498.44
Fine Crystalline Powder

Chemical Identifiers

6-3-6019
MFCD00150899
(-)-Nicotine di-(+)-hydrogen tartrate dihydrate
O.O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.CN1CCC[C@H]1C1=CN=CC=C1
498.44
LDMPZNTVIGIREC-HYRPJEHINA-N
N-{3-[(2Z)-5-chloro-2,3-dihydro-1,3-benzoxazol-2-ylidene]-4-oxocyclohexa-1,5-dien-1-yl}-2-methylpropanamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic to aquatic life with long lasting effects.
Fatal in contact with skin.
Fatal if inhaled.

GHS P Statement
IF ON SKIN: Gently wash with plenty of soap and water.
Wear protective gloves/protective clothing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only