missing translation for 'onlineSavingsMsg'
Learn More

Nicotinamide 99.0+%, TCI America™

Catalog No. N007825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N007825G 25 g
N0078500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N007825G Supplier TCI America Supplier No. N007825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98-92-0
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.127
MDL Number MFCD00006395
InChI Key DFPAKSUCGFBDDF-UHFFFAOYSA-N
Synonym nicotinamide, niacinamide, 3-pyridinecarboxamide, vitamin pp, nicotinic acid amide, papulex, aminicotin, amixicotyn, nicobion, nicotylamide
PubChem CID 936
ChEBI CHEBI:17154
IUPAC Name pyridine-3-carboxamide
SMILES C1=CC(=CN=C1)C(=O)N

Specifications

Melting Point 129°C
Color White
pH 6.0 to 7.5 (50g/L H2O soln.)
Quantity 25 g
Formula Weight 122.13
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nicotinamide
EINECSNumber (5)-0736
RTECSNumber QS3675000
TSCA Yes