missing translation for 'onlineSavingsMsg'
Learn More

Nicotinamide Impurity C, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111023000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111023000 Supplier Merck Emd Millipore Supplier No. PHR2088100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 1452-77-3
Melting Point 110°C (decomposition) (literature)
Molecular Formula C6H6N2O
Linear Formula C6H6N2O
MDL Number MFCD00023483
Quantity 100 mg
Synonym Picolinamide; 2-Pyridinecarboxamide
Molecular Weight (g/mol) 122.12
Formula Weight 122.12
Grade Certified Reference Material
Physical Form Powder
Show More Show Less