missing translation for 'onlineSavingsMsg'
Learn More

Nickelous Carbonate, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02003451
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02003451 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02003451 Supplier Avantor Supplier No. 276401

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 3333-67-3
Physical Form Powder
Quantity 500 g
Molecular Formula CNiO3
MDL Number MFCD00011144
Synonym nickel carbonate, nickelous carbonate, nickel ii carbonate, nickel monocarbonate, unii-wm50squ829, carbonic acid, nickel 2+ salt 1:1, nickel 2+ carbonate nico3, nickel 2+ ion carbonate, nickel carbonate nico3
InChI Key ZULUUIKRFGGGTL-UHFFFAOYSA-L
SMILES [Ni++].[O-]C([O-])=O
IUPAC Name nickel(2+);carbonate
Molecular Weight (g/mol) 118.70
PubChem CID 18746
Formula Weight 118.7
Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Chemical Name or Material Nickelous Carbonate
Show More Show Less