missing translation for 'onlineSavingsMsg'
Learn More

Nickelocene 98.0+%, TCI America™

Catalog No. D15741G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15741G 1 g
D15745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D15741G Supplier TCI America Supplier No. D15741G

Chemical Identifiers

CAS 1271-28-9
Molecular Formula C10H10Ni
Molecular Weight (g/mol) 188.883
MDL Number MFCD00001441
InChI Key RQKPFSQDBAZFJV-UHFFFAOYSA-N
Synonym bis cyclopentadienyl nickel ii
PubChem CID 24942185
IUPAC Name cyclopenta-1,3-diene;nickel(2+)
SMILES C1C=CC=[C-]1.C1C=CC=[C-]1.[Ni+2]

Specifications

Melting Point 173°C
Color Black-Green
Quantity 1 g
UN Number 1325
Formula Weight 188.88
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nickelocene
RTECSNumber QR6500000
TSCA Yes