missing translation for 'onlineSavingsMsg'
Learn More

Nickel tin oxide dihydrate

CAS: 12035-38-0 | NiO3Sn | 225.40 g/mol

Supplier:  Thermo Scientific Chemicals 03945236

 View more versions of this product

Catalog No. AA3945236


Only null left
Explore available promotions

Description

Description

It is used as an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Nickel tin oxide dihydrate
12035-38-0
500 g
MFCD00054011
Insoluble in water.
225.40
Odorless
Green
Powder
NiO3Sn
Nickel stannate
SHULAURIUQUJKN-UHFFFAOYSA-N
261.43 (225.40 Anhydrous)

Chemical Identifiers

12035-38-0
225.40
SHULAURIUQUJKN-UHFFFAOYSA-N
NiO3Sn
MFCD00054011
Nickel stannate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H350-H372-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H350i-H372

EINECSNumber : 234-824-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only