missing translation for 'onlineSavingsMsg'
Learn More

Nickel Sulfate Hexahydrate (Crystalline/Technical), Fisher Chemical

Catalog No. N723
Change view
Click to view available options
Quantity:
3 kg
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
N723 3 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N723 Supplier Thermo Fisher Scientific Supplier No. N72 3

Chemical Identifiers

CAS 10101-97-0
Molecular Formula H12NiO10S
Molecular Weight (g/mol) 262.839
MDL Number MFCD00149813
InChI Key RRIWRJBSCGCBID-UHFFFAOYSA-L
Synonym nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928
PubChem CID 5284429
ChEBI CHEBI:53437
IUPAC Name nickel(2+);sulfate;hexahydrate
SMILES O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]

Specifications

Melting Point 53°C
Color Blue Green
pH 4 to 6
Physical Form Solid
Quantity 3 kg
Formula Weight 262.85
Grade Technical
Identification Pass Test
Packaging Glass Bottle
Decomposition Information >280°C
Chemical Name or Material Nickel Sulfate Hexahydrate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Cancer hazard. Harmful by inhalation and if swallowed. Irritating to eyes and skin. May cause allergic respiratory and skin reaction. May cause irritation of respiratory tract. May cause harm to the unborn child. Possible risks of irreversible effects. Danger of serious damage to health by prolonged exposure. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention.

NFPA

Health:2
Flammability:1
Instability:0