missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Sulfate, Hexahydrate, Crystal, Purified, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Physical Form | Crystals and/or Chunks |
| Quantity | 45 kg |
| Molecular Weight (g/mol) | 262.85 |
| Grade | Purified |
| Packaging | Poly Bottle |
| Chemical Name or Material | Nickel Sulfate, Hexahydrate |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.