missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Sulfate, 0.05 Molar Standardized, Reagents
Specifications
Specifications
| Chemical Name or Material | Nickel Sulfate, 0.05 Molar Standardized |
| Boiling Point | 100°C |
| Color | Blue to Green |
| CAS | 7732-18-5, 10101-97-0 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Synonym | Nickelous Sulfate, Nickel (II) Sulfate |
| Grade | Reagent |
| Packaging | Poly Bottle |
| Specific Gravity | 1.0069 |