missing translation for 'onlineSavingsMsg'
Learn More

Nickel silicide, 99% (metals basis excluding Co), Co 0.1-1%, Thermo Scientific Chemicals

Catalog No. p-7050235
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3625118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3625118 Supplier Thermo Scientific Chemicals Supplier No. 03625118
Only null left

Ni silicides have been widely investigated for contact materials in microelectronic devices. Electroplating is another major use of the metal. nickel silicide/silicon nanowire heterostructures have been used to produce field-effect transistors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12059-14-2
Molecular Formula Ni2Si
Molecular Weight (g/mol) 145.472
MDL Number MFCD00151367
InChI Key RUFLMLWJRZAWLJ-UHFFFAOYSA-N
Synonym nickel silicide, dinickelylidenesilane
PubChem CID 14767304
SMILES [Si](=[Ni])=[Ni]

Specifications

Physical Form Powder
Quantity 50 g
UN Number UN3077
Solubility Information Insoluble in water.
Formula Weight 145.47
Percent Purity 99%
Odor Odorless
Assay (metals basis excluding Co), Co 0.1 to 1%
Chemical Name or Material Nickel silicide
Hazard Category H317-H350i-H372
Hazard Statement GHS H Statement
H350-H372-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 235-033-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only