missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel plating solution, semi-bright finish
$73.42 - $134.68
Specifications
| Chemical Name or Material | Nickel plating solution |
|---|---|
| Color | Green |
| Name Note | Semi-bright finish |
| Form | Liquid |
| MDL Number | MFCD00011137 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Nickel plating solution | |
| Semi-bright finish | |
| MFCD00011137 |
| Green | |
| Liquid | |
| UN3287 |
RUO – Research Use Only